Benzanthrone ≥98.0%
Supplier: TCI America
Synonyms:
7H-Benzo[de]anthracen-7-one
B0019-25G
B0019-500G
TCB0019-025GEA
75.84
CAD
TCB0019-025G
TCB0019-500G
Benzanthrone ≥98.0%
Benzanthrone
CAS Number: 82-05-3
MDL Number: MFCD00003585
Molecular Formula: C17H10O
Molecular Weight: 230.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 176
Formula:
C₁₇H₁₀O MW: 230.27 g/mol Melting Pt: 176 °C |
MDL Number:
MFCD00003585 CAS Number: 82-05-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...