Saccharin ≥99.0% for synthesis

Supplier: TCI America

Synonyms: o-Sulphobenzimide, o-Benzoic sulphimide, o-Benzoic sulfimide, o-Sulfobenzimide

B0004-25G B0004-500G
TCB0004-025GEA 29.55 CAD
TCB0004-025G TCB0004-500G
Saccharin ≥99.0% for synthesis
Saccharin

CAS Number: 81-07-2
MDL Number: MFCD00005866
Molecular Formula: C7H5NO3S
Molecular Weight: 183.18
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 226


Product Categories: Bioscience, Other Plant Hormones
Other Product Names / Synonyms: o-Benzoic Sulfimide, Saccharin

Formula: C₇H₅NO₃S
MW: 183.19 g/mol
Melting Pt: 226 °C
MDL Number: MFCD00005866
CAS Number: 81-07-2
EINECS: 201-321-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2ba26305cb16073ba88ab7c5af69ea59904449d&vid=d2ba26305cb16073ba88ab7c5af69ea59904449d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR