Dihexylammonium acetate ∼0.5 mol/L in water
Supplier: TCI America
A5705-10ML
A5705-100ML
TCA5705-10MLEA
63.42
CAD
TCA5705-10ML
TCA5705-100ML
Dihexylammonium acetate ∼0.5 mol/L in water
Dihexylammonium acetate
[Ion-Pair Reagent for LC-MS]
CAS Number: 366793-17-1
MDL Number: MFCD03093619
Molecular Formula: C14H31NO2
Molecular Weight: 245.41
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: IPC-DHAA
Formula:
C₁₄H₃₁NO₂ Density: 1.00 (20 °C) |
MDL Number:
MFCD03093619 CAS Number: 366793-17-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...