Diamylammonium acetate ∼0.5 mol/L in water

Supplier: TCI America

A5704-10ML A5704-100ML
TCA5704-10MLEA 71.1 CAD
TCA5704-10ML TCA5704-100ML
Diamylammonium acetate ∼0.5 mol/L in water
Diamylammonium acetate

[Ion-Pair Reagent for LC-MS]
CAS Number: 211676-91-4
MDL Number: MFCD03093618
Molecular Formula: C12H27NO2
Molecular Weight: 217.35
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: IPC-DAAA

Formula: C₁₂H₂₇NO₂
MDL Number: MFCD03093618
CAS Number: 211676-91-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7dd7b03522d5d03ef89acc133b2ab8cade2a26c8&vid=db532c616222d86d85d89bddcc99254457105464&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR