Dibutylammonium acetate ∼0.5 mol/L in water
Supplier: TCI America
A5702-10ML
A5702-100ML
TCA5702-10MLEA
53.04
CAD
TCA5702-10ML
TCA5702-100ML
Dibutylammonium acetate ∼0.5 mol/L in water
Dibutylammonium acetate
[Ion-Pair Reagent for LC-MS]
CAS Number: 19070-91-8
MDL Number: MFCD00191381
Molecular Formula: C8H19N
Molecular Weight: 189.30
Form: Clear Liquid
Color: Colorless
Other Product Names / Synonyms: IPC-DBAA
Formula:
C₈H₁₉N |
MDL Number:
MFCD00191381 CAS Number: 19070-91-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...