Sodium-9,10-dimethoxyanthracene-2-sulfonate 98,0

Supplier: TCI America

Synonyms: IPA-DAS, 9,10-Dimethoxyanthracene-2-sulphonic acid Sodium Salt, 9,10-Dimethoxyanthracene-2-sulfonic acid Sodium Salt

A5701-1G A5701-5G
TCA5701-001GEA 298.62 CAD
TCA5701-001G TCA5701-5G
Sodium-9,10-dimethoxyanthracene-2-sulfonate 98,0
Sodium-9,10-dimethoxyanthracene-2-sulfonate

[Fluorimetric Ion-Pair Reagent for Amines]
CAS Number: 67580-39-6
MDL Number: MFCD00042061
Molecular Formula: C16H14O5S
Molecular Weight: 340.32
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Analytical Science, Fluorimetric Ion-Pair Reagent for HPLC
Other Product Names / Synonyms: 9,10-Dimethoxyanthracene-2-sulfonic Acid Sodium Salt, IPA-DAS

Formula: C₁₆H₁₄O₅S
MW: 340.32 g/mol
MDL Number: MFCD00042061
CAS Number: 67580-39-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09f11b5dabf18afb7cc9b423cb6613803f21f3cf&vid=09f11b5dabf18afb7cc9b423cb6613803f21f3cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR