NAM [=N-(9-Acridinyl)maleimide] ≥98.0% (by HPLC)

Supplier: TCI America

A5591-50MG A5591-100MG
TCA5591-050MGEA 278.08 CAD
TCA5591-050MG TCA5591-100MG
NAM [=N-(9-Acridinyl)maleimide] ≥98.0% (by HPLC)
NAM [=N-(9-Acridinyl)maleimide]

[for HPLC Labeling]
CAS Number: 49759-20-8
MDL Number: MFCD00042983
Molecular Formula: C17H10N2O2
Molecular Weight: 274.28
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Bioscience, Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes, Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: N-(9-Acridinyl)maleimide, 9-Maleimidoacridine

Formula: C₁₇H₁₀N₂O₂
MDL Number: MFCD00042983
CAS Number: 49759-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df2fa4fbe8420f831cc5f55504ba452e41310e12&vid=df2fa4fbe8420f831cc5f55504ba452e41310e12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR