2-Amino-3-hydroxyanthraquinone ≥98.0%

Supplier: TCI America

A0315-500G A0315-25G
TCA0315-500GEA 1079.3 CAD
TCA0315-500G TCA0315-025G
2-Amino-3-hydroxyanthraquinone ≥98.0%
2-Amino-3-hydroxyanthraquinone

CAS Number: 117-77-1
MDL Number: MFCD00059499
Molecular Formula: C14H9NO3
Molecular Weight: 239.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₁₄H₉NO₃
MDL Number: MFCD00059499
CAS Number: 117-77-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0666d16c919d80aa3dd06b5cd6aacb08895c7a5d&vid=0666d16c919d80aa3dd06b5cd6aacb08895c7a5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR