Zinc(II) acetylacetonate ≥96.0% for synthesis
Supplier: TCI America
Synonyms:
Bis(2,4-pentanedionato-O,O')zinc (II), Zinc acetylacetonate, Zinc (II) 2,4-pentanedionate, Zinc (II) bis(2,4-pentanedionato-O,O')
Z0002-25G
Z0002-500G
Z0002-100G
TCZ0002-025GEA
120.4
CAD
TCZ0002-025G
TCZ0002-500G
TCZ0002-100G
Zinc(II) acetylacetonate ≥96.0% for synthesis
Zinc (II) acetylacetonate
CAS Number: 14024-63-6
MDL Number: MFCD00000035
Molecular Formula: C10H14O4Zn
Molecular Weight: 263.60
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: White
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts
Other Product Names / Synonyms: Acetylacetone Zinc(II) Salt, Zinc(II) Acetylacetonate
Formula:
C₁₀H₁₄O₄Zn MW: 263.61 g/mol |
MDL Number:
MFCD00000035 CAS Number: 14024-63-6 EINECS: 237-860-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...