Xanthane hydride ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: 5-Amino-3H-1,2,4-dithiazole-3-thione

X0001-25G X0001-500G X0001-100G
TCX0001-025GEA 213.44 CAD
TCX0001-025G TCX0001-500G TCX0001-100G
Xanthane hydride ≥95.0% (by HPLC)
Xanthane hydride

CAS Number: 6846-35-1
MDL Number: MFCD00051660
Molecular Formula: C2H2N2S3
Molecular Weight: 150.23
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: Pale Yellow

Formula: C₂H₂N₂S₃
MW: 150.23 g/mol
MDL Number: MFCD00051660
CAS Number: 6846-35-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7468519355575c0829ccc058be29b7e301ed0bf&vid=c7468519355575c0829ccc058be29b7e301ed0bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR