N-Vinylformamide ≥96.0% stabilised

Supplier: TCI America

V0106-25G V0106-500G
TCV0106-25GEA 43.37 CAD
TCV0106-25G TCV0106-500G
N-Vinylformamide ≥96.0% stabilised
N-Vinylformamide

(stabilized with BHT)
CAS Number: 13162-05-5
MDL Number: MFCD00081206
Molecular Formula: C3H5NO
Molecular Weight: 71.08
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 87
Melting point (°C): -16
Flash Point (°C): 106
Specific Gravity (20/20): 1.02
Lambda max.: 225 nm (MeOH)Storage Temperature: 0-10°C

Formula: C₃H₅NO
MW: 71.08 g/mol
Boiling Pt: 87 °C
Melting Pt: –16 °C
Density: 1.02 (20 °C)
Flash Pt: 106 °C
MDL Number: MFCD00081206
CAS Number: 13162-05-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73768ca341d55ab45e3a8a0191bb393fe24ba2cd&vid=73768ca341d55ab45e3a8a0191bb393fe24ba2cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR