L-Valinamide hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: H-Val-NH2.HCl, H-Val-NH₂·HCl

V0102-5G V0102-25G
TCV0102-5GEA 47.4 CAD
TCV0102-5G TCV0102-25G
L-Valinamide hydrochloride ≥98.0% (by total nitrogen and titration analysis)
L-Valinamide hydrochloride

CAS Number: 3014-80-0
MDL Number: MFCD00039085
Molecular Formula: C5H12N2O
Molecular Weight: 152.62
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Color: White
Melting point (°C): 263
Specific rotation [a]20/D: 27 deg (C=1, H2O)

Formula: C₅H₁₂N₂O
MW: 152.62 g/mol
Melting Pt: 263 °C
MDL Number: MFCD00039085
CAS Number: 3014-80-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83bf911fedd98c60406cf36b2d710fa2da968cf0&vid=83bf911fedd98c60406cf36b2d710fa2da968cf0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR