4-Vinylbenzylamine ≥92.0% stabilized

Supplier: TCI America

V0101-1G V0101-5G
TCV0101-1GEA 219.62 CAD
TCV0101-1G TCV0101-5G
4-Vinylbenzylamine ≥92.0% stabilized
4-Vinylbenzylamine

(stabilized with MEHQ)
CAS Number: 50325-49-0
MDL Number: MFCD08704374
Molecular Formula: C9H11N
Molecular Weight: 133.19
Purity/Analysis Method: >92.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 60
Specific Gravity (20/20): 0.98
Storage Temperature: <0°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4-(Aminomethyl)styrene

Formula: C₉H₁₁N
Boiling Pt: 60 °C
Density: 0.98 (20 °C)
MDL Number: MFCD08704374
CAS Number: 50325-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fda556dd027adfd78644067d9fa95d85c235f121&vid=fda556dd027adfd78644067d9fa95d85c235f121&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 92.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR