Agmatine sulfate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
1-Amino-4-guanidobutane Sulphate, 1-(4-Aminobutyl)guanidine sulfate, 1-(4-Aminobutyl)guanidine sulphate, 1-Amino-4-guanidobutane sulfate
A0310-1G
A0310-100MG
TCA0310-001GEA
77.64
CAD
TCA0310-001G
TCA0310-100MG
Agmatine sulfate ≥98.0% (by titrimetric analysis)
Agmatine sulfate
CAS Number: 2482-00-0
MDL Number: MFCD00013109
Molecular Formula: C5H14N4
Molecular Weight: 228.27
Purity/Analysis Method:
Other Product Names / Synonyms: 1-Amino-4-guanidobutane Sulfate
Formula:
C₅H₁₄N₄ MW: 228.27 g/mol Melting Pt: 231 °C |
MDL Number:
MFCD00013109 CAS Number: 2482-00-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...