DL-Valinol 98,0

Supplier: TCI America

Synonyms: 2-Amino-3-methylbutan-1-ol, DL-2-Amino-3-methyl-1-butanol, 2-Amino-3-methyl-1-butanol

V0100-5G V0100-1G
TCV0100-5GEA 249.64 CAD
TCV0100-5G TCV0100-1G
DL-Valinol 98,0
DL-Valinol

CAS Number: 16369-05-4
MDL Number: MFCD00004730
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 87
Flash Point (°C): 90


Other Product Names / Synonyms: 2-Amino-3-methyl-1-butanol

Formula: C₅H₁₃NO
MW: 103.17 g/mol
Boiling Pt: 87 °C
Flash Pt: 90 °C
MDL Number: MFCD00004730
CAS Number: 16369-05-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=253b09e69796ec630538ebd793d22cc82ca76111&vid=253b09e69796ec630538ebd793d22cc82ca76111&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR