D-Valine benzyl ester-p-toluenesulfonate ≥98.0%

Supplier: TCI America

Synonyms: H-D-Val-OBzl.Tos-OH, D-Valine Benzyl Ester p-toluenesulphonate, D-Valine Benzyl Ester p-Toluenesulfonate, H-D-Val-Obzl.TosOH

V0095-1G V0095-5G
TCV0095-1GEA 48.98 CAD
TCV0095-1G TCV0095-5G
D-Valine benzyl ester-p-toluenesulfonate ≥98.0%
D-Valine benzyl ester-p-toluenesulfonate

CAS Number: 17662-84-9
MDL Number: MFCD00136545
Molecular Formula: C12H17NO2
Molecular Weight: 379.47
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 162
Specific rotation [a]20/D: 4 deg (C=3.2, MeOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Val-OBzl.Tos-OH

Formula: C₁₂H₁₇NO₂
MW: 379.47 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00136545
CAS Number: 17662-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5915d39f97fab3d8dc789a822262da0f3238ff9&vid=b5915d39f97fab3d8dc789a822262da0f3238ff9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR