Vinyltris(2-methoxyethoxy)silane ≥96.0%

Supplier: TCI America

V0048-25ML V0048-500ML
TCV0048-025MLEA 52.14 CAD
TCV0048-025ML TCV0048-500ML
Vinyltris(2-methoxyethoxy)silane ≥96.0%
Vinyltris(2-methoxyethoxy)silane

CAS Number: 1067-53-4
MDL Number: MFCD00008500
Molecular Formula: C11H24O6Si
Molecular Weight: 280.39
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 113
Flash Point (°C): 132
Specific Gravity (20/20): 1.04


Product Categories: Synthetic Organic Chemistry, Vinylation, Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Tris(2-methoxyethoxy)vinylsilane

Formula: C₁₁H₂₄O₆Si
Boiling Pt: 113 °C
Density: 1.04 (20 °C)
Flash Pt: 132 °C
MDL Number: MFCD00008500
CAS Number: 1067-53-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ce4e712fa71102564526d050ad952aafa966536&vid=1ce4e712fa71102564526d050ad952aafa966536&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR