Undecafluoropentyl Iodide ≥98.0% stabilised

Supplier: TCI America

Synonyms: Perfluoropentyl Iodide

U0081-25G U0081-5G
TCU0081-25GEA 649.54 CAD
TCU0081-25G TCU0081-5G
Undecafluoropentyl Iodide ≥98.0% stabilised
Undecafluoropentyl Iodide

(stabilized with Copper chip)
CAS Number: 638-79-9
MDL Number: MFCD08461629
Molecular Formula: C5F11I
Molecular Weight: 395.94
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 94
Melting point (°C): -50
Specific Gravity (20/20): 2.06


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Perfluoropentyl Iodide

Formula: C₅F₁₁I
MW: 395.94 g/mol
Boiling Pt: 94 °C
Melting Pt: –50 °C
Density: 2.06 (20 °C)
MDL Number: MFCD08461629
CAS Number: 638-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c192df63172e028031c5056fb1b1b0a014d1136&vid=4c192df63172e028031c5056fb1b1b0a014d1136&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR