Undecafluoroiodocyclohexane ≥98.0% stabilized

Supplier: TCI America

U0080-1G U0080-5G
TCU0080-1GEA 74.26 CAD
TCU0080-1G TCU0080-5G
Undecafluoroiodocyclohexane ≥98.0% stabilized
Undecafluoroiodocyclohexane

(stabilized with Copper chip)
CAS Number: 355-69-1
MDL Number: MFCD00156095
Molecular Formula: C6F11I
Molecular Weight: 407.95
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 112
Specific Gravity (20/20): 2.18


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds
Other Product Names / Synonyms: Perfluoroiodocyclohexane

Formula: C₆F₁₁I
Boiling Pt: 112 °C
Density: 2.18 (20 °C)
MDL Number: MFCD00156095
CAS Number: 355-69-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=570f8b907864bb0f6c6cbd4b622d9af1d3b3a774&vid=570f8b907864bb0f6c6cbd4b622d9af1d3b3a774&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR