Ursolic acid ≥90.0%

Supplier: TCI America

U0065-1G U0065-100MG
TCU0065-001GEA 590.92 CAD
TCU0065-001G TCU0065-100MG
Ursolic acid ≥90.0%
Ursolic acid

CAS Number: 77-52-1
MDL Number: MFCD00009621
Molecular Formula: C30H48O3
Molecular Weight: 456.71
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Color: White
Specific rotation [a]20/D: 61 deg (C=1, Pyridine)


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Triterpenes
Other Product Names / Synonyms: 3beta-Hydroxy-12-ursen-28-oic Acid

Formula: C₃₀H₄₈O₃
MDL Number: MFCD00009621
CAS Number: 77-52-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8456eba824948e187bc8d9abf404b41d1cc4d27e&vid=8456eba824948e187bc8d9abf404b41d1cc4d27e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR