1,3,5-Undecatriene (mixture of isomers) ≥75.0%
Supplier: TCI America
U0062-5ML
U0062-25ML
TCU0062-005MLEA
186.44
CAD
TCU0062-005ML
TCU0062-25ML
1,3,5-Undecatriene (mixture of isomers) ≥75.0%
1,3,5-Undecatriene (mixture of isomers)
CAS Number: 16356-11-9
MDL Number: MFCD00015699
Molecular Formula: C11H18
Molecular Weight: 150.27
Purity/Analysis Method: >75.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 88
Flash Point (°C): 65
Specific Gravity (20/20): 0.80
Storage Temperature: 0-10°C
Formula:
C₁₁H₁₈ Boiling Pt: 88 °C Density: 0.80 (20 °C) Flash Pt: 65 °C |
MDL Number:
MFCD00015699 CAS Number: 16356-11-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...