2-Undecylimidazole ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Undecyl-1H-imidazole

U0012-500G U0012-25G
TCU0012-500GEA 508.76 CAD
TCU0012-500G TCU0012-025G
2-Undecylimidazole ≥96.0% (by titrimetric analysis)
2-Undecylimidazole

CAS Number: 16731-68-3
MDL Number: MFCD00038694
Molecular Formula: C14H26N2
Molecular Weight: 222.38
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 215
Melting point (°C): 73

Formula: C₁₄H₂₆N₂
MW: 222.3696 g/mol
Boiling Pt: 215 °C
Melting Pt: 73 °C
MDL Number: MFCD00038694
CAS Number: 16731-68-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8336628fecb7f3c01344ca8a9dd2d8c92e9be3eb&vid=8336628fecb7f3c01344ca8a9dd2d8c92e9be3eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR