3,3,5-Trimethylcyclohexanol ≥80.0% (contains ca. 20% trans- isomer)
Supplier: TCI America
T2693-25G
T2693-400G
TCT2693-25GEA
79
CAD
TCT2693-25G
TCT2693-400G
3,3,5-Trimethylcyclohexanol ≥80.0% (contains ca. 20% trans- isomer)
3,3,5-Trimethylcyclohexanol
CAS Number: 116-02-9
MDL Number: MFCD00019378
Molecular Formula: C9H18O
Molecular Weight: 142.24
Purity/Analysis Method: >80.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 86
Melting point (°C): 34
Flash Point (°C): 88
Formula:
C₉H₁₈O Boiling Pt: 86 °C Melting Pt: 34 °C Density: 0.9006 g/cm³ (16 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019378 CAS Number: 116-02-9 EINECS: 204-122-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...