Tris(2-pyridylmethyl)amine ≥98.0%

Supplier: TCI America

T2671-1G T2671-5G
TCT2671-1GEA 340.73 CAD
TCT2671-1G TCT2671-5G
Tris(2-pyridylmethyl)amine ≥98.0%
Tris(2-pyridylmethyl)amine

CAS Number: 16858-01-8
Molecular Formula: C18H18N4
Molecular Weight: 290.37
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 86

Formula: C₁₈H₁₈N₄
Melting Pt: 86 °C
MDL Number: MFCD14708176
CAS Number: 16858-01-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e47de75b869438ff35ce239a60b3e9ac1231e076&vid=e47de75b869438ff35ce239a60b3e9ac1231e076&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR