1,3,5-Tris(4-carboxyphenyl)benzene ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 5'-(4-Carboxyphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid

T2647-1G T2647-5G
TCT2647-1GEA 218.04 CAD
TCT2647-1G TCT2647-5G
1,3,5-Tris(4-carboxyphenyl)benzene ≥98.0% (by HPLC)
1,3,5-Tris(4-carboxyphenyl)benzene

CAS Number: 50446-44-1
MDL Number: MFCD10000888
Molecular Formula: C27H18O6
Molecular Weight: 438.44
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 325

Formula: C₂₇H₁₈O₆
MW: 438.44 g/mol
Melting Pt: 325 °C
MDL Number: MFCD10000888
CAS Number: 50446-44-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c4c6470ef26cd7c6a44f261b2172540bed14a66&vid=4c4c6470ef26cd7c6a44f261b2172540bed14a66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR