4,4,5,5-Tetramethyl-1,3,2-dioxaphospholane-2-oxide ≥95.0%
Supplier: TCI America
T2645-1G
T2645-5G
TCT2645-1GEA
153.26
CAD
TCT2645-1G
TCT2645-5G
4,4,5,5-Tetramethyl-1,3,2-dioxaphospholane-2-oxide ≥95.0%
4,4,5,5-Tetramethyl-1,3,2-dioxaphospholane-2-oxide
CAS Number: 16352-18-4
MDL Number: MFCD09836231
Molecular Formula: C6H13O3P
Molecular Weight: 164.14
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 104
Formula:
C₆H₁₃O₃P Melting Pt: 104 °C |
MDL Number:
MFCD09836231 CAS Number: 16352-18-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...