1,3,5-Tris(4-bromophenyl)benzene ≥98.0%
Supplier: TCI America
Synonyms:
4,4''-Dibromo-5'-(4-bromophenyl)-1,1':3',1''-terphenyl
T2644-1G
T2644-5G
TCT2644-1GEA
82.16
CAD
TCT2644-1G
TCT2644-5G
1,3,5-Tris(4-bromophenyl)benzene ≥98.0%
1,3,5-Tris(4-bromophenyl)benzene
CAS Number: 7511-49-1
MDL Number: MFCD00362911
Molecular Formula: C24H15Br3
Molecular Weight: 543.10
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 263
Lambda max.: 288 nm (CHCl3)
Formula:
C₂₄H₁₅Br₃ MW: 543.10 g/mol Melting Pt: 263 °C |
MDL Number:
MFCD00362911 CAS Number: 7511-49-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...