Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate ≥97.0%

Supplier: TCI America

Synonyms: DOTA(OtBu)₃, 2-(4,7,10-Tris(2-(tert-butoxy)-2-oxoethyl)-1,4,7,10-tetraazacyclododecan-1-yl)acetic acid, 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic Acid Tri-tert-butyl Ester, 4,7,10-Tri-(t-butyloxycarbonylmethyl)-1,4,7,10-tetraazacyclododecan-1-yl-acetic acid hydrobromide salt, DOTA-tri(t-butyl ester)

T2642-100MG T2642-1G
TCT2642-100MGEA 262.28 CAD
TCT2642-100MG TCT2642-1G
Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate ≥97.0%
Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate

CAS Number: 137076-54-1
MDL Number: MFCD02259697
Molecular Formula: C28H52N4O8
Molecular Weight: 572.74
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic Acid Tri-tert-butyl Ester

Formula: C₂₈H₅₂N₄O₈
MW: 572.74 g/mol
MDL Number: MFCD02259697
CAS Number: 137076-54-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e07095627992f8810686f3593808ba0c247f6ac&vid=0e07095627992f8810686f3593808ba0c247f6ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR