1,3,5-Tri-O-benzoyl-ɑ-D-ribofuranose ≥97.0%
Supplier: TCI America
Synonyms:
1,3,5-Tri-O-benzoyl-α-D-ribofuranose, 1,3,5-Tri-O-benzoyl-a-D-ribofuranose
T2641-5G
T2641-25G
TCT2641-5GEA
158.63
CAD
TCT2641-5G
TCT2641-25G
1,3,5-Tri-O-benzoyl-ɑ-D-ribofuranose ≥97.0%
1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose
CAS Number: 22224-41-5
MDL Number: MFCD00080818
Molecular Formula: C26H22O8
Molecular Weight: 462.45
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 140
Specific rotation [a]20/D: 85 deg (C=1, CHCl3)
Lambda max.: 273 nm (MeOH)
Formula:
C₂₆H₂₂O₈ MW: 462.45 g/mol Melting Pt: 140 °C |
MDL Number:
MFCD00080818 CAS Number: 22224-41-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...