2,4,6-Triphenylboroxin ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
T2640-25G
T2640-5G
TCT2640-25GEA
459.78
CAD
TCT2640-25G
TCT2640-5G
2,4,6-Triphenylboroxin ≥98.0% (by HPLC, titration analysis)
2,4,6-Triphenylboroxin
CAS Number: 3262-89-3
MDL Number: MFCD00014587
Molecular Formula: C18H15B3O3
Molecular Weight: 311.75
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 219
Formula:
C₁₈H₁₅B₃O₃ Melting Pt: 219 °C |
MDL Number:
MFCD00014587 CAS Number: 3262-89-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...