2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione ≥98.0%

Supplier: TCI America

Synonyms: Hajos-Parrish Ketone

T2635-1G T2635-5G
TCT2635-1GEA 203.35 CAD
TCT2635-1G TCT2635-5G
2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione ≥98.0%
2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione

CAS Number: 19576-08-0
MDL Number: MFCD00135278
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 71


Other Product Names / Synonyms: Hajos-Parrish Ketone

Formula: C₁₀H₁₂O₂
MW: 164.20 g/mol
Melting Pt: 71 °C
MDL Number: MFCD00135278
CAS Number: 19576-08-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98fc2f642a61be4d02543b4ecfe8ca520606f93c&vid=98fc2f642a61be4d02543b4ecfe8ca520606f93c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR