Triethylene glycol-2-bromoethyl methyl ether ≥95.0%

Supplier: TCI America

T2634-5G T2634-25G
TCT2634-5GEA 294.2 CAD
TCT2634-5G TCT2634-25G
Triethylene glycol-2-bromoethyl methyl ether ≥95.0%
Triethylene glycol-2-bromoethyl methyl ether

CAS Number: 110429-45-3
Molecular Formula: C9H19BrO4
Molecular Weight: 271.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 1.29
Storage Temperature: 0-10°C


Product Categories: Bioscience, PEGylation Reagents & PEG Spacers, PEGylation Reagents
Other Product Names / Synonyms: 2-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]ethyl Bromide, 1-Bromo-3,6,9,12-tetraoxatridecane, mPEG4-Bromide

Formula: C₉H₁₉BrO₄
Density: 1.29 (20 °C)
MDL Number: MFCD12545963
CAS Number: 110429-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aaee1e11d03295711680ecd894f2293beee77546&vid=aaee1e11d03295711680ecd894f2293beee77546&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR