1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: 3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, Togni Reagent

T2624-1G T2624-5G
TCT2624-1GEA 347.13 CAD
TCT2624-1G TCT2624-5G
1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole ≥97.0% (by HPLC)
1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole

CAS Number: 887144-97-0
MDL Number: MFCD10567056
Molecular Formula: C10H10F3IO
Molecular Weight: 330.09
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 78
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Hypervalent Iodine Compounds, Fluorinating Agents, Building Blocks, Fluorinated Bioactive Compounds
Other Product Names / Synonyms: Togni Reagent

Formula: C₁₀H₁₀F₃IO
MW: 330.09 g/mol
Melting Pt: 78 °C
MDL Number: MFCD10567056
CAS Number: 887144-97-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b18ca522e21b9ca18dc3121cb895d7f6786b468&vid=7b18ca522e21b9ca18dc3121cb895d7f6786b468&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR