Thiopicolinamide ≥97.0%

Supplier: TCI America

Synonyms: Pyridine-2-carbothioamide, 2-Pyridinethioamide, 2-Pyridylthiocarboxamide

T2618-5G T2618-1G
TCT2618-5GEA 109.02 CAD
TCT2618-5G TCT2618-1G
Thiopicolinamide ≥97.0%
Thiopicolinamide

CAS Number: 5346-38-3
MDL Number: MFCD00087576
Molecular Formula: C6H6N2S
Molecular Weight: 138.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 138
Flash Point (°C): 160


Other Product Names / Synonyms: 2-Pyridylthiocarboxamide

Formula: C₆H₆N₂S
Melting Pt: 138 °C
Flash Pt: 160 °C
MDL Number: MFCD00087576
CAS Number: 5346-38-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eab8ce3e462781fb63de21a968d200bde6280434&vid=eab8ce3e462781fb63de21a968d200bde6280434&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR