4,4',4''-Tri-9-carbazolyltriphenylamine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Tris(4-carbazol-9-ylphenyl)amine, 4,4',4''-Tris(carbazol-9-yl)triphenylamine, Tris(4-(9H-carbazol-9-yl)phenyl)amine, TCTA

T2616-1G T2616-5G
TCT2616-1GEA 107.44 CAD
TCT2616-1G TCT2616-5G
4,4',4''-Tri-9-carbazolyltriphenylamine ≥98.0% (by HPLC, total nitrogen)
4,4',4''-Tri-9-carbazolyltriphenylamine

CAS Number: 139092-78-7
MDL Number: MFCD03093250
Molecular Formula: C54H36N4
Molecular Weight: 740.91
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 266
Lambda max.: 326 nm (CH2Cl2)


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials
Other Product Names / Synonyms: TCTA

Formula: C₅₄H₃₆N₄
MW: 740.91 g/mol
Melting Pt: 266 °C
MDL Number: MFCD03093250
CAS Number: 139092-78-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf82263f6fe27ff92e8954660cad0507f26eea44&vid=bf82263f6fe27ff92e8954660cad0507f26eea44&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR