Tetraphenylphosphonium tetra-p-tolylborate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T2603-25G T2603-5G
TCT2603-25GEA 555.37 CAD
TCT2603-25G TCT2603-5G
Tetraphenylphosphonium tetra-p-tolylborate ≥98.0% (by HPLC, titration analysis)
Tetraphenylphosphonium tetra-p-tolylborate

CAS Number: 181259-35-8
MDL Number: MFCD08274957
Molecular Formula: C52H48BP
Molecular Weight: 714.74
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White

Formula: C₅₂H₄₈BP
MDL Number: MFCD08274957
CAS Number: 181259-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e86b40315ac686c1f6586fecf91611fc262c0f5a&vid=e86b40315ac686c1f6586fecf91611fc262c0f5a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR