2-Hydroxy-3-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T2600-1G
T2600-5G
TCT2600-1GEA
66.36
CAD
TCT2600-1G
TCT2600-5G
2-Hydroxy-3-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
2-Hydroxy-3-(trifluoromethyl)pyridine
CAS Number: 22245-83-6
MDL Number: MFCD00153194
Molecular Formula: C6H4F3NO
Molecular Weight: 163.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 155
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(Trifluoromethyl)-2-pyridinone, 2-Hydroxy-3-(trifluoromethyl)pyridine
Formula:
C₆H₄F₃NO Melting Pt: 155 °C |
MDL Number:
MFCD00153194 CAS Number: 22245-83-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...