2,2',7,7'-Tetrabromo-9,9'-spirobifluorene ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T2597-1G T2597-5G
TCT2597-1GEA 97.96 CAD
TCT2597-1G TCT2597-5G
2,2',7,7'-Tetrabromo-9,9'-spirobifluorene ≥98.0% (by titrimetric analysis)
2,2',7,7'-Tetrabromo-9,9'-spirobifluorene

CAS Number: 128055-74-3
MDL Number: MFCD08704220
Molecular Formula: C25H12Br4
Molecular Weight: 631.99
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₂₅H₁₂Br₄
MDL Number: MFCD08704220
CAS Number: 128055-74-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dbbf8ecb7a2cc55891c3142e8953e0f1ce45502&vid=9dbbf8ecb7a2cc55891c3142e8953e0f1ce45502&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR