1-[2-(Trimethylsilyl)ethoxycarbonyloxy]benzotriazole ≥98.0%

Supplier: TCI America

Synonyms: Teoc-OBt

T2590-1G T2590-5G
TCT2590-1GEA 260.7 CAD
TCT2590-1G TCT2590-5G
1-[2-(Trimethylsilyl)ethoxycarbonyloxy]benzotriazole ≥98.0%
1-[2-(Trimethylsilyl)ethoxycarbonyloxy]benzotriazole

CAS Number: 113306-55-1
Molecular Formula: C12H17N3O3Si
Molecular Weight: 279.37
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: Teoc-OBt

Formula: C₁₂H₁₇N₃O₃Si
MW: 279.37 g/mol
CAS Number: 113306-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efeb52d7350cbac9c135c2c425602457c4a64bad&vid=efeb52d7350cbac9c135c2c425602457c4a64bad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR