(+)-Terpinen-4-ol ≥93.0%

Supplier: TCI America

T2581-5G T2581-25G
TCT2581-5GEA 64.69 CAD
TCT2581-5G TCT2581-25G
(+)-Terpinen-4-ol ≥93.0%
(+)-Terpinen-4-ol

CAS Number: 2438-10-0
MDL Number: MFCD00077431
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 90
Flash Point (°C): 79
Specific Gravity (20/20): 0.94
Specific rotation [a]20/D: 20 deg (neat)


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: (+)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol, (+)-4-Hydroxy-4-isopropyl-1-methyl-1-cyclohexene, (+)-p-Menth-1-en-4-ol

Formula: C₁₀H₁₈O
Boiling Pt: 90 °C
Density: 0.94 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00077431
CAS Number: 2438-10-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a32310c4af5754c7218b8f72d1fc0457bdb975e&vid=1a32310c4af5754c7218b8f72d1fc0457bdb975e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR