2,3,4,6-Tetra-O-acetyl-D-mannopyranosyl fluoride ≥95.0%

Supplier: TCI America

T2567-1G T2567-5G
TCT2567-1GEA 396.74 CAD
TCT2567-1G TCT2567-5G
2,3,4,6-Tetra-O-acetyl-D-mannopyranosyl fluoride ≥95.0%
2,3,4,6-Tetra-O-acetyl-D-mannopyranosyl fluoride

CAS Number: 174511-17-2
Molecular Formula: C14H19FO9
Molecular Weight: 350.30
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Storage Temperature: 0-10°C

Formula: C₁₄H₁₉FO₉
MW: 350.30 g/mol
CAS Number: 174511-17-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4049b359c27915eb0cb7897cdde8f54e1404566e&vid=4049b359c27915eb0cb7897cdde8f54e1404566e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR