(Triphenylphosphoranylidene)ketene

Supplier: TCI America

T2565-1G T2565-5G
TCT2565-1GEA 173.8 CAD
TCT2565-1G TCT2565-5G
(Triphenylphosphoranylidene)ketene
(Triphenylphosphoranylidene)ketene

CAS Number: 15596-07-3
MDL Number: MFCD00040613
Molecular Formula: C20H15OP
Molecular Weight: 302.31
Form: Crystal
Color: White
Melting point (°C): 171


Other Product Names / Synonyms: Ketenylidene(triphenyl)phosphorane, Bestmann Ylide

Formula: C₂₀H₁₅OP
Melting Pt: 171 °C
MDL Number: MFCD00040613
CAS Number: 15596-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9effb0ed4c1277dfdad2162aede07c7041896f4&vid=e9effb0ed4c1277dfdad2162aede07c7041896f4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR