Tetraethyl-4-tert-butylcalix[4]arene-O,O',O'',O'''-tetraacetate ≥96.0% (by HPLC)

Supplier: TCI America

T2563-200MG T2563-1G
TCT2563-200MGEA 246.48 CAD
TCT2563-200MG TCT2563-1G
Tetraethyl-4-tert-butylcalix[4]arene-O,O',O'',O'''-tetraacetate ≥96.0% (by HPLC)
Tetraethyl-4-tert-butylcalix[4]arene-O,O',O'',O'''-tetraacetate

CAS Number: 97600-39-0
MDL Number: MFCD00145373
Molecular Formula: C60H80O12
Molecular Weight: 993.29
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 155


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 4-tert-Butylcalix[4]arene-O,O',O'',O'''-tetraacetic Acid Tetraethyl Ester

Formula: C₆₀H₈₀O₁₂
Melting Pt: 155 °C
MDL Number: MFCD00145373
CAS Number: 97600-39-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d05591959d5b3e49a34a32c3e59144a130809079&vid=d05591959d5b3e49a34a32c3e59144a130809079&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR