2,3-Dihydro-1H-carbazol-4(9H)-one ≥98.0%

Supplier: TCI America

Synonyms: Ondansetron Impurity I, 1,2,3,4-Tetrahydro-4-oxocarbazole, 1,2,3,4-Tetrahydrocarbazol-4-one

T2559-5G T2559-25G
TCT2559-5GEA 28.44 CAD
TCT2559-5G TCT2559-25G
2,3-Dihydro-1H-carbazol-4(9H)-one ≥98.0%
2,3-Dihydro-1H-carbazol-4(9H)-one

CAS Number: 15128-52-6
MDL Number: MFCD00173749
Molecular Formula: C12H11NO
Molecular Weight: 185.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 226


Other Product Names / Synonyms: 1,2,3,4-Tetrahydro-4-oxocarbazole

Formula: C₁₂H₁₁NO
MW: 185.2218 g/mol
Melting Pt: 226 °C
MDL Number: MFCD00173749
CAS Number: 15128-52-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd593debaec443ce747674ed6fc6a1b6e2e04925&vid=fd593debaec443ce747674ed6fc6a1b6e2e04925&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR