Tri-p-tolylamine ≥98.0%

Supplier: TCI America

T2558-25G T2558-5G
TCT2558-25GEA 668.34 CAD
TCT2558-25G TCT2558-5G
Tri-p-tolylamine ≥98.0%
Tri-p-tolylamine

CAS Number: 1159-53-1
MDL Number: MFCD00674043
Molecular Formula: C21H21N
Molecular Weight: 287.41
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 170
Melting point (°C): 117

Formula: C₂₁H₂₁N
Boiling Pt: 170 °C
Melting Pt: 117 °C
MDL Number: MFCD00674043
CAS Number: 1159-53-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c160b7c4db5e762d3ce77ad971b8d2cb893cf5e9&vid=c160b7c4db5e762d3ce77ad971b8d2cb893cf5e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR