2',3',5'-Tri-O-acetyluridine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3,4-diyl diacetate, Uridine triacetate, Uridine 2',3',5'-triacetate

T2549-25G T2549-5G
TCT2549-25GEA 161.16 CAD
TCT2549-25G TCT2549-5G
2',3',5'-Tri-O-acetyluridine ≥98.0% (by HPLC, titration analysis)
2',3',5'-Tri-O-acetyluridine

CAS Number: 4105-38-8
MDL Number: MFCD00023795
Molecular Formula: C15H18N2O9
Molecular Weight: 370.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 129


Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: Uridine 2',3',5'-Triacetate

Formula: C₁₅H₁₈N₂O₉
MW: 370.31 g/mol
Melting Pt: 129 °C
MDL Number: MFCD00023795
CAS Number: 4105-38-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af9878ae36c80ed220e4d5fad4e6149fe6320518&vid=af9878ae36c80ed220e4d5fad4e6149fe6320518&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR