3-Bromomethyl-7-methoxy-1,4-benzoxazin-2-one ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A5553-1G A5553-100MG
TCA5553-001GEA 692.51 CAD
TCA5553-001G TCA5553-100MG
3-Bromomethyl-7-methoxy-1,4-benzoxazin-2-one ≥98.0% (by titrimetric analysis)
3-Bromomethyl-7-methoxy-1,4-benzoxazin-2-one

[for HPLC Labeling]
CAS Number: 124522-09-4
MDL Number: MFCD00143099
Molecular Formula: C10H8BrNO3
Molecular Weight: 270.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: 0-10°C

Formula: C₁₀H₈BrNO₃
MDL Number: MFCD00143099
CAS Number: 124522-09-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=707bc5acd56c5e43a8c13040eae6178a48574184&vid=707bc5acd56c5e43a8c13040eae6178a48574184&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR