(R)-(p-Toluenesulfinyl)ferrocene ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T2545-1G T2545-5G
TCT2545-1GEA 633.58 CAD
TCT2545-1G TCT2545-5G
(R)-(p-Toluenesulfinyl)ferrocene ≥98.0% (by HPLC, titration analysis)
(R)-(p-Toluenesulfinyl)ferrocene

CAS Number: 130225-27-3
Molecular Formula: C17H16FeOS
Molecular Weight: 324.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 143
Specific rotation [a]20/D: -400 deg (C=0.1, EtOAc)


Product Categories: Synthetic Organic Chemistry, Sulfoxides
Other Product Names / Synonyms: (R)-Ferrocenyl p-Tolyl Sulfoxide

Formula: C₁₇H₁₆FeOS
Melting Pt: 143 °C
CAS Number: 130225-27-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=710ba3dd262bca580ebed8d92e04e5916600d633&vid=710ba3dd262bca580ebed8d92e04e5916600d633&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR