2-(Trimethylsilyl)ethyl-3-Nitro-1H-1,2,4-triazole-1-carboxylate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T2544-5G T2544-1G
TCT2544-5GEA 766.3 CAD
TCT2544-5G TCT2544-1G
2-(Trimethylsilyl)ethyl-3-Nitro-1H-1,2,4-triazole-1-carboxylate ≥98.0% (by titrimetric analysis)
2-(Trimethylsilyl)ethyl-3-Nitro-1H-1,2,4-triazole-1-carboxylate

CAS Number: 1001067-09-9
Molecular Formula: C8H14N4O4Si
Molecular Weight: 258.31
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: 3-Nitro-1H-1,2,4-triazole-1-carboxylic Acid 2-(Trimethylsilyl)ethyl Ester, 3-Nitro-1-[2-(trimethylsilyl)ethoxycarbonyl]-1H-1,2,4-triazole, Teoc-NT

Formula: C₈H₁₄N₄O₄Si
MDL Number: MFCD11042686
CAS Number: 1001067-09-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18343d721bd54f2aa023986f62caf1a14cb3e11c&vid=18343d721bd54f2aa023986f62caf1a14cb3e11c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR