2,3,5,6-Tetrafluoro-4-mercaptobenzoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

T2542-5G T2542-1G
TCT2542-5GEA 905.34 CAD
TCT2542-5G TCT2542-1G
2,3,5,6-Tetrafluoro-4-mercaptobenzoic acid ≥97.0% (by GC, titration analysis)
2,3,5,6-Tetrafluoro-4-mercaptobenzoic acid

CAS Number: 5211-44-9
Molecular Formula: C7H2F4O2S
Molecular Weight: 226.14
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 153

Formula: C₇H₂F₄O₂S
Melting Pt: 153 °C
CAS Number: 5211-44-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=269d6a68013569f99634717dbe1cc1492fc5f0a1&vid=269d6a68013569f99634717dbe1cc1492fc5f0a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR